Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILF3
Entrez Gene ID
3609
Gene Name
interleukin enhancer binding factor 3
Gene Aliases
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80
Chromosome Location
Chr.19: 10654261 - 10692419 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137673.1 NP_001131145.1 3 - 4 550 66
NM_004516.3 NP_004507.2 3 - 4 550 66
NM_012218.3 NP_036350.2 3 - 4 550 66
NM_017620.2 NP_060090.2 3 - 4 550 66
NM_153464.2 NP_703194.1 3 - 4 550 66
XM_006722742.2 XP_006722805.1 3 - 4 465 66
XM_011527984.2 XP_011526286.2 3 - 4 583 66
XM_011527985.2 XP_011526287.1 3 - 4 465 66
XM_011527987.1 XP_011526289.1 1 - 2 34 66
XM_017026763.1 XP_016882252.1 4 - 5 674 66
XM_017026764.1 XP_016882253.1 1 - 2 230 66
XM_017026765.1 XP_016882254.1 3 - 4 465 66
GenBank mRNA AB209062.1 - 3 - 4 507 66
AF141870.1 - 3 - 4 344 66
AF147209.1 - 1 - 2 245 66
AF167569.1 - 1 - 2 233 66
AF167570.1 - 1 - 2 233 66
AJ271745.1 - 2 - 3 322 66
AJ271746.1 - 2 - 3 302 66
AK291205.1 - 3 - 4 449 66
AK291617.1 - 3 - 4 435 66
AK294084.1 - 3 - 4 441 66
BC001770.1 - 3 - 4 435 66
BC048314.1 - 3 - 4 480 66
BC064836.1 - 3 - 4 483 66
HQ322263.1 - 1 - 2 233 66
HQ322265.1 - 1 - 2 233 66
X98264.1 - 3 - 4 437 66
X98265.1 - 2 - 3 442 66

View Full Product Details