The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144823.2 | NP_001138295.1 | 10 - 11 | 1693 | 64 |
NM_001320835.1 | NP_001307764.1 | 10 - 11 | 1693 | 64 | |
NM_005848.4 | NP_005839.3 | 10 - 11 | 1686 | 64 | |
XM_005254120.4 | XP_005254177.1 | 9 - 10 | 1472 | 64 | |
XM_005254121.3 | XP_005254178.1 | 10 - 11 | 1693 | 64 | |
XM_005254122.4 | XP_005254179.1 | 10 - 11 | 1552 | 64 | |
XM_006720371.3 | XP_006720434.1 | 10 - 11 | 1428 | 64 | |
XM_011521156.2 | XP_011519458.1 | 10 - 11 | 1688 | 64 | |
XM_017021862.1 | XP_016877351.1 | 9 - 10 | 1413 | 64 | |
XM_017021863.1 | XP_016877352.1 | 9 - 10 | 1471 | 64 | |
XM_017021864.1 | XP_016877353.1 | 10 - 11 | 1688 | 64 | |
XM_017021865.1 | XP_016877354.1 | 10 - 11 | 1551 | 64 | |
GenBank mRNA | AF534403.1 | - | 10 - 11 | 1648 | 64 |
AK290292.1 | - | 10 - 11 | 1501 | 64 | |
AK299570.1 | - | 10 - 11 | 1607 | 64 | |
AK307411.1 | - | 10 - 11 | 1693 | 64 | |
AL832602.2 | - | 8 - 9 | 1212 | 64 | |
AL833317.1 | - | 10 - 11 | 1524 | 64 | |
AL833384.1 | - | 7 - 8 | 957 | 64 | |
BC028057.1 | - | 10 - 11 | 1538 | 64 | |
BC041706.1 | - | 10 - 11 | 1517 | 64 |