The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286102.1 | NP_001273031.1 | 5 - 6 | 763 | 63 |
NM_003269.4 | NP_003260.1 | 5 - 6 | 1356 | 63 | |
GenBank mRNA | AB307709.1 | - | 5 - 6 | 648 | 63 |
AF220532.1 | - | 5 - 6 | 1355 | 63 | |
AF411525.1 | - | 5 - 6 | 889 | 63 | |
AK131541.1 | - | 5 - 6 | 763 | 63 | |
BC028031.1 | - | 5 - 6 | 853 | 63 | |
HQ692846.1 | - | 5 - 6 | 648 | 63 | |
Y13276.1 | - | 5 - 6 | 816 | 63 |