The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286102.1 | NP_001273031.1 | 7 - 8 | 1001 | 100 |
NM_003269.4 | NP_003260.1 | 7 - 8 | 1594 | 100 | |
GenBank mRNA | AB307709.1 | - | 7 - 8 | 886 | 100 |
AF220532.1 | - | 7 - 8 | 1593 | 100 | |
AF411525.1 | - | 7 - 8 | 1127 | 100 | |
AK131541.1 | - | 7 - 8 | 1001 | 100 | |
BC028031.1 | - | 7 - 8 | 1091 | 100 | |
HQ692846.1 | - | 7 - 8 | 886 | 100 | |
Y13276.1 | - | 7 - 8 | 1054 | 100 |