Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
Chromosome Location
Chr.5: 169637247 - 170083382 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004946.2 NP_004937.1 29 - 30 3066 68
GenBank mRNA AK128107.1 - 15 - 16 1627 68
BC104900.1 - 29 - 30 3022 68
BC113457.1 - 29 - 30 3022 68
BC143358.1 - 29 - 30 3022 68
D86964.1 - 29 - 30 3024 68

View Full Product Details