The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004946.2 | NP_004937.1 | 3 - 4 | 243 | 99 |
XM_005265830.3 | XP_005265887.1 | 3 - 4 | 243 | 99 | |
XM_011534448.2 | XP_011532750.1 | 3 - 4 | 243 | 99 | |
XM_011534449.2 | XP_011532751.1 | 3 - 4 | 243 | 99 | |
XM_011534450.2 | XP_011532752.1 | 3 - 4 | 243 | 99 | |
XM_011534451.2 | XP_011532753.1 | 3 - 4 | 243 | 99 | |
XM_017009189.1 | XP_016864678.1 | 3 - 4 | 243 | 99 | |
XM_017009190.1 | XP_016864679.1 | 3 - 4 | 243 | 99 | |
GenBank mRNA | BC104900.1 | - | 3 - 4 | 199 | 99 |
BC113457.1 | - | 3 - 4 | 199 | 99 | |
BC143358.1 | - | 3 - 4 | 199 | 99 | |
D86964.1 | - | 3 - 4 | 201 | 99 | |
DB145764.1 | - | 3 - 4 | 243 | 99 |