Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOTCH3
Entrez Gene ID
4854
Gene Name
notch 3
Gene Aliases
CADASIL, CADASIL1, CASIL, IMF2, LMNS
Chromosome Location
Chr.19: 15159633 - 15200981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000435.2 NP_000426.2 8 - 9 1454 70
XM_005259924.4 XP_005259981.1 8 - 9 1393 70
GenBank mRNA AB209447.1 - 8 - 9 1387 70
U97669.1 - 8 - 9 1456 70

View Full Product Details