Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WARS2
Entrez Gene ID
10352
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Gene Aliases
TrpRS
Chromosome Location
Chr.1: 119031216 - 119140701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015836.3 NP_056651.1 5 - 6 659 106
XM_005270350.3 XP_005270407.1 5 - 6 745 106
XM_011540494.2 XP_011538796.1 6 - 7 728 106
XM_011540495.2 XP_011538797.1 4 - 5 430 106
XM_017000039.1 XP_016855528.1 7 - 8 914 106
XM_017000040.1 XP_016855529.1 5 - 6 481 106
XM_017000042.1 XP_016855531.1 4 - 5 565 106
GenBank mRNA AJ242739.1 - 5 - 6 640 106
AK223197.1 - 5 - 6 637 106
AK296799.1 - 4 - 5 554 106
AK299547.1 - 5 - 6 598 106
AK300295.1 - 5 - 6 487 106
AK313740.1 - 5 - 6 657 106
BC044575.1 - 5 - 6 642 106

View Full Product Details