Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDST3
Entrez Gene ID
9348
Gene Name
N-deacetylase and N-sulfotransferase 3
Gene Aliases
HSST3
Chromosome Location
Chr.4: 118033596 - 118258945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004784.2 NP_004775.1 10 - 11 2523 96
XM_006714416.3 XP_006714479.1 10 - 11 2579 96
XM_017008839.1 XP_016864328.1 10 - 11 2491 96
XM_017008840.1 XP_016864329.1 10 - 11 2361 96
XM_017008841.1 XP_016864330.1 9 - 10 4202 96
XM_017008842.1 XP_016864331.1 9 - 10 1806 96
XM_017008843.1 XP_016864332.1 7 - 8 984 96
XM_017008844.1 XP_016864333.1 9 - 10 1241 96
GenBank mRNA AF074924.1 - 10 - 11 2523 96
AF076605.1 - 9 - 10 2309 96
BC109309.1 - 10 - 11 2370 96
BC109310.1 - 10 - 11 2370 96

View Full Product Details