The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004784.2 | NP_004775.1 | 12 - 13 | 2798 | 111 |
XM_006714416.3 | XP_006714479.1 | 12 - 13 | 2854 | 111 | |
XM_017008839.1 | XP_016864328.1 | 12 - 13 | 2766 | 111 | |
XM_017008840.1 | XP_016864329.1 | 12 - 13 | 2636 | 111 | |
XM_017008841.1 | XP_016864330.1 | 11 - 12 | 4477 | 111 | |
XM_017008842.1 | XP_016864331.1 | 11 - 12 | 2081 | 111 | |
XM_017008843.1 | XP_016864332.1 | 9 - 10 | 1259 | 111 | |
XM_017008844.1 | XP_016864333.1 | 11 - 12 | 1516 | 111 | |
GenBank mRNA | AF074924.1 | - | 12 - 13 | 2798 | 111 |
AF076605.1 | - | 11 - 12 | 2584 | 111 | |
BC109309.1 | - | 12 - 13 | 2645 | 111 | |
BC109310.1 | - | 12 - 13 | 2645 | 111 |