Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDST3
Entrez Gene ID
9348
Gene Name
N-deacetylase and N-sulfotransferase 3
Gene Aliases
HSST3
Chromosome Location
Chr.4: 118033596 - 118258945 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004784.2 NP_004775.1 4 - 5 1629 67
XM_006714416.3 XP_006714479.1 4 - 5 1685 67
XM_011532415.2 XP_011530717.1 4 - 5 1597 67
XM_011532416.2 XP_011530718.1 4 - 5 1597 67
XM_017008839.1 XP_016864328.1 4 - 5 1597 67
XM_017008840.1 XP_016864329.1 4 - 5 1467 67
XM_017008841.1 XP_016864330.1 3 - 4 3308 67
XM_017008842.1 XP_016864331.1 3 - 4 912 67
XM_017008844.1 XP_016864333.1 3 - 4 347 67
GenBank mRNA AF074924.1 - 4 - 5 1629 67
AF076605.1 - 3 - 4 1415 67
BC109309.1 - 4 - 5 1476 67
BC109310.1 - 4 - 5 1476 67

View Full Product Details