The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267721.1 | NP_001254650.1 | 4 - 5 | 785 | 114 |
NM_144603.3 | NP_653204.1 | 4 - 5 | 770 | 114 | |
NM_172167.2 | NP_751907.1 | 4 - 5 | 773 | 114 | |
NM_172168.2 | NP_751908.1 | 4 - 5 | 788 | 114 | |
XM_005255099.4 | XP_005255156.1 | 5 - 6 | 1883 | 114 | |
XM_017022927.1 | XP_016878416.1 | 5 - 6 | 1389 | 114 | |
XM_017022928.1 | XP_016878417.1 | 5 - 6 | 746 | 114 | |
GenBank mRNA | AB097667.1 | - | 4 - 5 | 583 | 114 |
AF532983.1 | - | 4 - 5 | 404 | 114 | |
AF532984.1 | - | 4 - 5 | 407 | 114 | |
AF532985.1 | - | 4 - 5 | 422 | 114 | |
AF539796.1 | - | 4 - 5 | 403 | 114 | |
AK292197.1 | - | 4 - 5 | 773 | 114 | |
AK310198.1 | - | 2 - 3 | 975 | 114 | |
AK313917.1 | - | 4 - 5 | 693 | 114 | |
AY191359.1 | - | 4 - 5 | 419 | 114 | |
AY255768.1 | - | 4 - 5 | 400 | 114 | |
BC015917.2 | - | 4 - 5 | 660 | 114 |