The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317770.1 | NP_001304699.1 | 2 - 3 | 402 | 80 |
NM_017592.2 | NP_060062.1 | 2 - 3 | 402 | 80 | |
GenBank mRNA | AK000411.1 | - | 2 - 3 | 312 | 80 |
AK124629.1 | - | 3 - 4 | 265 | 80 | |
AK294667.1 | - | 3 - 4 | 224 | 80 | |
AK297772.1 | - | 2 - 3 | 293 | 80 | |
AK298014.1 | - | 2 - 3 | 312 | 80 | |
AK300563.1 | - | 2 - 3 | 394 | 80 | |
AK316010.1 | - | 3 - 4 | 233 | 80 | |
AL137304.1 | - | 2 - 3 | 317 | 80 | |
AY729650.1 | - | 2 - 3 | 382 | 80 | |
BC019015.2 | - | 2 - 3 | 309 | 80 |