The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317781.1 | NP_001304710.1 | 18 - 19 | 2736 | 67 |
XM_005263267.4 | XP_005263324.1 | 18 - 19 | 3179 | 67 | |
XM_011532302.2 | XP_011530604.1 | 20 - 21 | 3314 | 67 | |
XM_011532303.2 | XP_011530605.1 | 20 - 21 | 3311 | 67 | |
XM_011532304.2 | XP_011530606.1 | 19 - 20 | 3278 | 67 | |
XM_011532305.2 | XP_011530607.1 | 19 - 20 | 3215 | 67 | |
XM_011532307.2 | XP_011530609.1 | 19 - 20 | 3149 | 67 | |
XM_011532308.2 | XP_011530610.1 | 18 - 19 | 3113 | 67 | |
XM_017008681.1 | XP_016864170.1 | 19 - 20 | 3149 | 67 | |
XM_017008682.1 | XP_016864171.1 | 17 - 18 | 3014 | 67 | |
XM_017008683.1 | XP_016864172.1 | 17 - 18 | 2223 | 67 | |
XM_017008684.1 | XP_016864173.1 | 19 - 20 | 2495 | 67 | |
XM_017008685.1 | XP_016864174.1 | 13 - 14 | 1941 | 67 | |
GenBank mRNA | AF113223.1 | - | 12 - 13 | 1621 | 67 |
AK095661.1 | - | 15 - 16 | 2140 | 67 | |
AY044656.1 | - | 11 - 12 | 1536 | 67 | |
AY601647.1 | - | 18 - 19 | 2736 | 67 | |
BC029868.1 | - | 12 - 13 | 1811 | 67 |