The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114357.1 | NP_001107829.1 | 5 - 6 | 739 | 173 |
XM_005263019.3 | XP_005263076.1 | 7 - 8 | 1092 | 173 | |
XM_011531989.2 | XP_011530291.1 | 6 - 7 | 1018 | 173 | |
XM_017008236.1 | XP_016863725.1 | 7 - 8 | 2485 | 173 | |
XM_017008237.1 | XP_016863726.1 | 8 - 9 | 1260 | 173 | |
XM_017008238.1 | XP_016863727.1 | 6 - 7 | 2269 | 173 | |
XM_017008239.1 | XP_016863728.1 | 6 - 7 | 2272 | 173 | |
XM_017008240.1 | XP_016863729.1 | 6 - 7 | 1021 | 173 | |
XM_017008241.1 | XP_016863730.1 | 5 - 6 | 2247 | 173 | |
GenBank mRNA | BC127739.1 | - | 3 - 4 | 444 | 173 |
BC141967.1 | - | 2 - 3 | 173 | ||
BC141968.1 | - | 3 - 4 | 443 | 173 | |
BC150574.1 | - | 5 - 6 | 739 | 173 |