Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3A
Entrez Gene ID
7380
Gene Name
uroplakin 3A
Gene Aliases
UP3A, UPIII, UPIIIA, UPK3
Chromosome Location
Chr.22: 45284987 - 45295874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006953.3 NP_008884.1 3 - 4 510 81
GenBank mRNA AB010116.1 - 2 - 3 387 81
AB010637.1 - 3 - 4 483 81
AF085808.1 - 3 - 4 515 81
BC069544.1 - 3 - 4 515 81
CR456608.1 - 3 - 4 485 81

View Full Product Details