Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CYP4V2 | NM_207352.3 | 11 | 3449 | NP_997235.3 |
XM_005262935.3 | 11 | 3231 | XP_005262992.1 | |
XM_017008037.1 | 10 | 3207 | XP_016863526.1 | |
AK022114.1 | 5 | 2332 | ||
AK126473.1 | 12 | 3118 | BAC86562.1 | |
BC041839.1 | 4 | 2883 | AAH41839.1 | |
BX648537.1 | 5 | 2380 | ||
BX648730.1 | 6 | 7203 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894090 | Chr4:186011626 - 186212421 on Build GRCh38 | Gain | CYP4V2 TLR3 FAM149A FLJ38576 |
nsv1028146 | Chr4:185582745 - 186417181 on Build GRCh38 | Gain | CYP4V2 SORBS2 TLR3 FAM149A KLKB1 FLJ38576 F11 F11-AS1 |
nsv4644 | Chr4:186185600 - 186219291 on Build GRCh38 | Insertion | CYP4V2 KLKB1 FLJ38576 |
nsv830172 | Chr4:186091611 - 186247270 on Build GRCh38 | Loss | CYP4V2 FAM149A KLKB1 FLJ38576 |
nsv508340 | Chr4:186138019 - 186215804 on Build GRCh38 | Deletion | CYP4V2 FAM149A KLKB1 FLJ38576 |
esv3603523 | Chr4:186176579 - 186214960 on Build GRCh38 | Gain | CYP4V2 FLJ38576 |
nsv461864 | Chr4:186085652 - 186335850 on Build GRCh38 | Gain | CYP4V2 TLR3 FAM149A KLKB1 FLJ38576 F11 F11-AS1 |
nsv596401 | Chr4:186151229 - 186445495 on Build GRCh38 | Loss | CYP4V2 FAM149A KLKB1 FLJ38576 F11 F11-AS1 |
esv3603517 | Chr4:186008703 - 186213065 on Build GRCh38 | Gain | CYP4V2 TLR3 FAM149A FLJ38576 |
dgv1052e214 | Chr4:186072002 - 186212619 on Build GRCh38 | Gain | CYP4V2 TLR3 FAM149A FLJ38576 |
nsv4642 | Chr4:186164028 - 186215260 on Build GRCh38 | Deletion | CYP4V2 FAM149A FLJ38576 |
esv3603525 | Chr4:186210472 - 186225457 on Build GRCh38 | Loss | CYP4V2 KLKB1 |