Product Details

Assay Reference Genome
Location

Chr.5:132660920 on build GRCh38
Cytoband
5q31.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3596
Gene Symbol
IL13
Gene Name
interleukin 13
Gene Aliases
IL-13, P600
Location
Chr.5:132658173-132661109 on build GRCh38
Assay Gene Location
Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IL13 NM_002188.2 4 1093 NP_002179.2
L06801.1 4 1081 AAA36107.1
X69079.1 4 1093 CAA48823.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv950392 Chr5:132655609 - 132670708 on Build GRCh38 Deletion TH2LCRR IL13
nsv528281 Chr5:132650771 - 132682252 on Build GRCh38 Gain TH2LCRR IL13 IL4 LOC105379176

View Full Product Details