Product Details

Assay Reference Genome
Location

Chr.7:100408604 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Overlaps Intron 10 - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK000919.1 BAA91424.1
AK093038.1 BAC04028.1
AK300745.1
AK302108.1
AL136735.1 CAB66669.1
BC002725.1 AAH02725.1
KU178677.1
KU178678.1
KU178679.1
KU178680.1
KU178681.1
KU178682.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion NYAP1 LAMTOR4 GATS GNB2 AP1S1 PCOLCE-AS1 ACTL6B UFSP1 CLDN15 TSC22D4 FIS1 PILRB TAF6 ZASP GAL3ST4 PLOD3 STAG3L5P-PVRIG2P-PILRB NAT16 TFR2 AZGP1 SAP25 AGFG2 PPP1R35 PVRIG TRIM56 SLC12A9 PILRA FBXO24 ZKSCAN1 CNPY4 STAG3 POP7 TRIP6 MUC17 SPDYE3 PVRIG2P MIR93 ZSCAN21 MIR25 MIR6875 MUC3A EPO MBLAC1 LOC105375429 MIR4653 MIR106B SRRT COPS6 C7orf43 MIR6840 AP4M1 LOC102724094 GIGYF1 ZNHIT1 EPHB4 MUC12 STAG3L5P ZAN VGF MOSPD3 ZCWPW1 PMS2P1 C7orf61 LRCH4 MIR4658 GPC2 PCOLCE MOGAT3 AZGP1P1 ZNF3 SERPINE1 ACHE MEPCE MCM7
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss NYAP1 MIR6840 ZCWPW1 PILRA PMS2P1 SPDYE3 PVRIG2P TSC22D4 PILRB C7orf61 STAG3L5P-PVRIG2P-PILRB STAG3L5P PPP1R35 MEPCE

View Full Product Details