Product Details

Assay Reference Genome
Location

Chr.7:30971116 on build GRCh38
Cytoband
7p14.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2692
Gene Symbol
GHRHR
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
Location
Chr.7:30964021-30979531 on build GRCh38
Assay Gene Location
Overlaps Intron 4 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GHRHR NM_000823.3 NP_000814.2
XM_011515263.1 XP_011513565.1
AF282259.1 AAG15368.1
AF282260.1 AAG15369.1
AF282261.1 AAG15370.1
AF282262.1 AAG15371.1
AY557192.1 AAS59864.1
L01406.1 AAA35890.1
L09237.1 AAA58619.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv6316n100 Chr7:30609596 - 31145482 on Build GRCh38 Gain FAM188B INMT-FAM188B INMT GARS CRHR2 ADCYAP1R1 GHRHR AQP1
nsv527038 Chr7:30617851 - 31122331 on Build GRCh38 Gain FAM188B INMT-FAM188B INMT GARS CRHR2 ADCYAP1R1 GHRHR AQP1
esv2761316 Chr7:30615466 - 31127062 on Build GRCh38 Gain FAM188B INMT-FAM188B INMT GARS CRHR2 ADCYAP1R1 GHRHR AQP1
nsv436557 Chr7:30288062 - 32730902 on Build GRCh38 Deletion PPP1R17 DPY19L1P1 INMT GARS CRHR2 ZNRF2P1 AQP1 ZNRF2 FAM188B MIR550B1 LOC401320 LOC100130673 AVL9 MIR550A1 LINC01176 DKFZP586I1420 PDE1C NEUROD6 GGCT INMT-FAM188B CCDC129 ADCYAP1R1 GHRHR NOD1 LOC101928268 LSM5

View Full Product Details