Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 16 - 17 1967 84
XM_011534141.2 XP_011532443.1 16 - 17 2049 84
XM_011534142.2 XP_011532444.1 16 - 17 1961 84
XM_011534143.2 XP_011532445.1 15 - 16 1923 84
XM_011534145.2 XP_011532447.1 10 - 11 1695 84
XM_011534146.2 XP_011532448.1 15 - 16 2528 84
GenBank mRNA AB051506.2 - 16 - 17 1953 84
AK024507.1 - 3 - 4 307 84

View Full Product Details