Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUC5B
Entrez Gene ID
727897
Gene Name
mucin 5B, oligomeric mucus/gel-forming
Gene Aliases
MG1, MUC-5B, MUC5, MUC9
Chromosome Location
Chr.11: 1223065 - 1262176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002458.2 NP_002449.2 41 - 42 16506 89
GenBank mRNA AK303892.1 - 8 - 9 1204 89
BM740816.1 - 3 - 4 235 89
U78551.1 - 4 - 5 331 89

View Full Product Details