Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
Chromosome Location
Chr.1: 167094048 - 167129165 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080426.1 NP_001073895.1 3 - 4 439 73
XM_005245603.3 XP_005245660.1 4 - 5 602 73
XM_011510146.2 XP_011508448.1 3 - 4 476 73
GenBank mRNA BC115387.1 - 4 - 5 559 73

View Full Product Details