The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024594.2 | NP_001019765.1 | 2 - 3 | 467 | 144 |
GenBank mRNA | BC038214.1 | - | 2 - 3 | 366 | 144 |
BC130608.1 | - | 2 - 3 | 467 | 144 | |
BC130634.1 | - | 2 - 3 | 467 | 144 |