The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265614.2 | NP_001252543.1 | 3 - 4 | 837 | 111 |
XM_005251688.1 | XP_005251745.1 | 3 - 4 | 592 | 111 | |
XM_006716948.2 | XP_006717011.1 | 3 - 4 | 993 | 111 | |
XM_006716949.3 | XP_006717012.1 | 3 - 4 | 853 | 111 | |
GenBank mRNA | AB067483.1 | - | 3 - 4 | 836 | 111 |