The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022831.2 | NP_073742.2 | 7 - 8 | 794 | 80 |
GenBank mRNA | AK022868.1 | - | 7 - 8 | 746 | 80 |
AK289865.1 | - | 7 - 8 | 745 | 80 | |
AK301870.1 | - | 7 - 8 | 920 | 80 | |
AL833718.1 | - | 7 - 8 | 584 | 80 | |
BC015535.1 | - | 7 - 8 | 721 | 80 | |
BC043142.1 | - | 7 - 8 | 1842 | 80 | |
BC067805.1 | - | 7 - 8 | 742 | 80 | |
BC171848.1 | - | 7 - 8 | 769 | 80 |