Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
Chromosome Location
Chr.22: 32413845 - 32464465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011530088.2 XP_011528390.1 1 - 2 92 69
XM_011530091.2 XP_011528393.1 1 - 2 92 69
XM_017028740.1 XP_016884229.1 1 - 2 92 69
GenBank mRNA AK301685.1 - 1 - 2 69 69

View Full Product Details