Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
Chromosome Location
Chr.22: 41699503 - 41799456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152513.3 NP_689726.3 7 - 8 896 67
XM_011529935.1 XP_011528237.1 7 - 8 905 67
XM_011529936.1 XP_011528238.1 7 - 8 905 67
XM_011529937.1 XP_011528239.1 7 - 8 905 67
XM_011529938.1 XP_011528240.1 7 - 8 905 67
XM_011529939.1 XP_011528241.1 7 - 8 905 67
XM_011529940.1 XP_011528242.1 7 - 8 905 67
XM_011529941.1 XP_011528243.1 7 - 8 905 67
XM_011529942.2 XP_011528244.1 7 - 8 1096 67
XM_011529943.1 XP_011528245.1 7 - 8 905 67
XM_011529944.1 XP_011528246.1 7 - 8 905 67
XM_011529945.2 XP_011528247.1 7 - 8 907 67
XM_011529946.1 XP_011528248.1 7 - 8 905 67
XM_011529947.1 XP_011528249.1 7 - 8 905 67
XM_011529948.2 XP_011528250.1 3 - 4 277 67
XM_011529949.1 XP_011528251.1 2 - 3 214 67
XM_011529950.1 XP_011528252.1 7 - 8 905 67
XM_011529952.1 XP_011528254.1 2 - 3 344 67
XM_011529955.1 XP_011528257.1 7 - 8 905 67
XM_011529956.2 XP_011528258.1 7 - 8 905 67
XM_017028633.1 XP_016884122.1 7 - 8 905 67
GenBank mRNA BC025400.1 - 7 - 8 912 67
BC035720.1 - 7 - 8 895 67
BC070111.1 - 2 - 3 346 67

View Full Product Details