Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB41
Entrez Gene ID
360023
Gene Name
zinc finger and BTB domain containing 41
Gene Aliases
FRBZ1, ZNF924
Chromosome Location
Chr.1: 197153682 - 197200542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194314.2 NP_919290.2 5 - 6 1738 102
NR_135153.1 5 - 6 1738 102
XM_011509507.2 XP_011507809.1 5 - 6 1717 102
XM_017001205.1 XP_016856694.1 5 - 6 1717 102
GenBank mRNA AK128549.1 - 5 - 6 1717 102
AY163816.2 - 5 - 6 1738 102
AY336933.1 - 2 - 3 209 102
BC132818.1 - 5 - 6 1702 102
BC136600.1 - 5 - 6 1702 102

View Full Product Details