Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RP1
- Entrez Gene ID
-
6101
- Gene Name
- retinitis pigmentosa 1 (autosomal dominant)
- Gene Aliases
-
DCDC4A, ORP1
- Chromosome Location
-
Chr.8: 54509448 - 54630834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK304443.1 |
- |
21 - 22 |
2640 |
89 |
AK316511.1 |
- |
21 - 22 |
3074 |
89 |
- Gene Symbol
- LOC107984125
- Entrez Gene ID
-
107984125
- Gene Name
- lipoxygenase homology domain-containing protein 1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 54657854 - 54871720 on Build GRCh38
View Full Product Details