The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143962.1 | NP_001137434.1 | 14 - 15 | 1723 | 66 |
XM_017001265.1 | XP_016856754.1 | 11 - 12 | 1164 | 66 | |
XM_017001266.1 | XP_016856755.1 | 9 - 10 | 1081 | 66 | |
GenBank mRNA | BC157893.1 | - | 14 - 15 | 1660 | 66 |