The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_037584.2 | 1 - 2 | 150 | 105 | |
GenBank mRNA | AF381171.1 | - | 1 - 2 | 116 | 105 |
AF381172.1 | - | 1 - 2 | 119 | 105 | |
AF381173.1 | - | 1 - 2 | 126 | 105 | |
AF381174.1 | - | 1 - 2 | 126 | 105 |