Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
-
Chromosome Location
Chr.9: 130265742 - 130434123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291815.1 NP_001278744.1 64 - 65 9825 76
XM_011518465.2 XP_011516767.1 64 - 65 9962 76
XM_011518466.2 XP_011516768.1 64 - 65 9962 76
XM_011518467.2 XP_011516769.1 63 - 64 9785 76
XM_011518468.2 XP_011516770.1 64 - 65 9962 76
XM_011518469.2 XP_011516771.1 64 - 65 9962 76
XM_017014585.1 XP_016870074.1 43 - 44 6631 76
XM_017014586.1 XP_016870075.1 18 - 19 2559 76

View Full Product Details