Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
-
Chromosome Location
Chr.9: 130265742 - 130434123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291815.1 NP_001278744.1 65 - 66 9952 86
XM_011518465.2 XP_011516767.1 65 - 66 10089 86
XM_011518466.2 XP_011516768.1 65 - 66 10089 86
XM_011518467.2 XP_011516769.1 64 - 65 9912 86
XM_011518468.2 XP_011516770.1 65 - 66 10089 86
XM_011518469.2 XP_011516771.1 65 - 66 10089 86
XM_017014585.1 XP_016870074.1 44 - 45 6758 86
XM_017014586.1 XP_016870075.1 19 - 20 2686 86
GenBank mRNA AL834139.1 - 6 - 7 665 86

View Full Product Details