Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MOXD2P
- Entrez Gene ID
-
100289017
- Gene Name
- monooxygenase, DBH-like 2, pseudogene
- Gene Aliases
-
MOXD2
- Chromosome Location
-
Chr.7: 142240737 - 142247067 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024346.3 |
|
7 - 8 |
|
114 |
GenBank mRNA |
DY655576.1 |
- |
4 - 5 |
310 |
114 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DY655575.1 |
- |
1 - 2 |
|
|
Chr. 7 - 142240774 - 142243403 [-] on Build GRCh38 |
114 |
GenBankmRNA |
DY654652.1 |
- |
4 - 5 |
308 |
|
Chr. 7 - 142240814 - 142243442 [+] on Build GRCh38 |
114 |
GenBankmRNA |
DY654834.1 |
- |
2 - 3 |
120 |
|
Chr. 7 - 142242569 - 142243784 [+] on Build GRCh38 |
114 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
maltase-glucoamylase 2 (putative) |
MGAM2 |
|
-19.3kb
|
Sense |
Chr. - 142111703 - 142223733 on Build GRCh38 |
Downstream |
protease, serine 58 |
PRSS58 |
PRSS1, TRY1, TRYX3, UNQ2540 |
8.7kb
|
Anti-sense |
Chr. - 142252138 - 142258058 on Build GRCh38 |
View Full Product Details