The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137675.3 | NP_001131147.1 | 1 - 2 | 114 | 71 |
GenBank mRNA | AB621810.1 | - | 1 - 2 | 129 | 71 |
DB097328.1 | - | 1 - 2 | 116 | 71 |