The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013641.2 | NP_001013663.1 | 1 - 2 | 151 | 69 |
GenBank mRNA | BC127706.1 | - | 1 - 2 | 131 | 69 |
BC127707.1 | - | 1 - 2 | 131 | 69 | |
BC137239.1 | - | 1 - 2 | 151 | 69 | |
BC137240.1 | - | 1 - 2 | 151 | 69 |