The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114091.2 | NP_001107563.1 | 1 - 2 | 167 | 59 |
NM_001256.4 | NP_001247.3 | 1 - 2 | 167 | 59 | |
NM_001293089.1 | NP_001280018.1 | 1 - 2 | 167 | 59 | |
NM_001293091.1 | NP_001280020.1 | 1 - 2 | 167 | 59 | |
XM_011525546.2 | XP_011523848.1 | 1 - 2 | 154 | 59 | |
XM_011525547.2 | XP_011523849.1 | 1 - 2 | 154 | 59 | |
XM_011525548.2 | XP_011523850.1 | 1 - 2 | 187 | 59 | |
XM_011525550.2 | XP_011523852.1 | 1 - 2 | 123 | 59 | |
XM_017025481.1 | XP_016880970.1 | 1 - 2 | 154 | 59 | |
XM_017025482.1 | XP_016880971.1 | 1 - 2 | 154 | 59 | |
XM_017025483.1 | XP_016880972.1 | 1 - 2 | 154 | 59 | |
XM_017025484.1 | XP_016880973.1 | 1 - 2 | 183 | 59 | |
XM_017025485.1 | XP_016880974.1 | 1 - 2 | 184 | 59 | |
XM_017025486.1 | XP_016880975.1 | 1 - 2 | 104 | 59 | |
XM_017025487.1 | XP_016880976.1 | 1 - 2 | 186 | 59 | |
GenBank mRNA | AK296881.1 | - | 1 - 2 | 107 | 59 |
AK299342.1 | - | 1 - 2 | 119 | 59 | |
AU135593.1 | - | 1 - 2 | 155 | 59 | |
BC011656.2 | - | 1 - 2 | 31 | 59 | |
BI548077.1 | - | - | 159 | 59 | |
BP234964.1 | - | 1 - 2 | 154 | 59 | |
HY023336.1 | - | 1 - 2 | 168 | 59 | |
S78234.1 | - | 1 - 2 | 127 | 59 | |
U00001.1 | - | 1 - 2 | 85 | 59 |