The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110822.1 | NP_001104292.1 | 6 - 7 | 882 | 120 |
XM_011527471.2 | XP_011525773.1 | 6 - 7 | 642 | 120 | |
XM_011527472.2 | XP_011525774.1 | 6 - 7 | 642 | 120 | |
XM_011527473.2 | XP_011525775.1 | 6 - 7 | 642 | 120 | |
XM_011527474.2 | XP_011525776.1 | 5 - 6 | 522 | 120 | |
XM_011527475.2 | XP_011525777.1 | 6 - 7 | 642 | 120 | |
XM_011527476.2 | XP_011525778.1 | 6 - 7 | 642 | 120 | |
XM_017027458.1 | XP_016882947.1 | 6 - 7 | 642 | 120 | |
GenBank mRNA | AB211063.1 | - | 5 - 6 | 564 | 120 |
BC049000.1 | - | 6 - 7 | 882 | 120 |