The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008743.2 | NP_001008743.1 | 5 - 6 | 623 | 109 |
NM_001320878.1 | NP_001307807.1 | 5 - 6 | 623 | 109 | |
XM_017004153.1 | XP_016859642.1 | 4 - 5 | 1512 | 109 | |
XM_017004154.1 | XP_016859643.1 | 4 - 5 | 1512 | 109 | |
XM_017004155.1 | XP_016859644.1 | 4 - 5 | 1512 | 109 |