The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039840.2 | NP_001034929.2 | 1 - 2 | 431 | 94 |
NM_001300884.1 | NP_001287813.1 | 1 - 2 | 431 | 94 | |
XM_017029582.1 | XP_016885071.1 | 1 - 2 | 534 | 94 | |
XM_017029584.1 | XP_016885073.1 | 1 - 2 | 534 | 94 | |
GenBank mRNA | AK295903.1 | - | 1 - 2 | 434 | 94 |
AK297768.1 | - | 1 - 2 | 288 | 94 | |
AL832095.1 | - | 1 - 2 | 71 | 94 | |
BC127727.1 | - | 1 - 2 | 342 | 94 | |
BC127728.1 | - | 1 - 2 | 342 | 94 | |
BC136691.1 | - | 1 - 2 | 336 | 94 | |
BC136692.1 | - | 1 - 2 | 336 | 94 | |
CR936642.1 | - | 1 - 2 | 382 | 94 | |
DC382882.1 | - | 1 - 2 | 432 | 94 |