The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177663.1 | NP_001171134.1 | 31 - 32 | 6903 | 118 |
NM_001177664.1 | NP_001171135.1 | 31 - 32 | 6806 | 118 | |
NM_001177665.1 | NP_001171136.1 | 30 - 31 | 7076 | 118 | |
GenBank mRNA | AY727910.1 | - | 31 - 32 | 6903 | 118 |
AY727911.1 | - | 31 - 32 | 6806 | 118 | |
AY727912.1 | - | 30 - 31 | 7076 | 118 | |
U79263.1 | - | 2 - 3 | 423 | 118 |