The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304840.1 | NP_001291769.1 | 14 - 15 | 3082 | 108 |
NM_020760.2 | NP_065811.1 | 16 - 17 | 3416 | 108 | |
XM_006712646.3 | XP_006712709.1 | 16 - 17 | 3474 | 108 | |
XM_006712648.3 | XP_006712711.1 | 15 - 16 | 3407 | 108 | |
XM_017004563.1 | XP_016860052.1 | 16 - 17 | 3453 | 108 | |
GenBank mRNA | AB037722.1 | - | 16 - 17 | 3416 | 108 |
BC117194.1 | - | 16 - 17 | 3332 | 108 | |
BC117198.1 | - | 16 - 17 | 3332 | 108 | |
CR749424.1 | - | 14 - 15 | 108 |