The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020760.2 | NP_065811.1 | 7 - 8 | 1070 | 70 |
XM_006712646.3 | XP_006712709.1 | 7 - 8 | 1107 | 70 | |
XM_006712648.3 | XP_006712711.1 | 6 - 7 | 1040 | 70 | |
XM_017004563.1 | XP_016860052.1 | 7 - 8 | 1107 | 70 | |
GenBank mRNA | AB037722.1 | - | 7 - 8 | 1070 | 70 |
BC117194.1 | - | 7 - 8 | 986 | 70 | |
BC117198.1 | - | 7 - 8 | 986 | 70 |