Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYW3
Entrez Gene ID
127253
Gene Name
tRNA-yW synthesizing protein 3 homolog
Gene Aliases
C1orf171
Chromosome Location
Chr.1: 74733143 - 74766677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138467.2 NP_612476.1 2 - 3 344 76
NR_027962.1 2 - 3 435 76
XM_006710347.1 XP_006710410.1 2 - 3 357 76
GenBank mRNA AK098237.1 - 2 - 3 344 76
AK311272.1 - 2 - 3 381 76
BC009514.1 - 2 - 3 278 76
BC071765.1 - 2 - 3 344 76

View Full Product Details