Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTX3
Entrez Gene ID
345778
Gene Name
metaxin 3
Gene Aliases
-
Chromosome Location
Chr.5: 79976716 - 79991267 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010891.4 NP_001010891.4 6 - 7 611 123
NM_001167741.1 NP_001161213.1 5 - 6 541 123
XM_005248495.3 XP_005248552.1 6 - 7 613 123
XM_017009440.1 XP_016864929.1 5 - 6 535 123
GenBank mRNA AK296864.1 - 6 - 7 606 123
BX538064.1 - 5 - 6 521 123

View Full Product Details