The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213656.3 | NP_998821.3 | 4 - 5 | 911 | 66 |
GenBank mRNA | AJ786657.1 | - | 4 - 5 | 911 | 66 |
BC157887.1 | - | 4 - 5 | 905 | 66 | |
BC171907.1 | - | 5 - 6 | 949 | 66 |