Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD19
Entrez Gene ID
149478
Gene Name
BTB domain containing 19
Gene Aliases
-
Chromosome Location
Chr.1: 44807442 - 44814199 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136537.1 NP_001130009.1 2 - 3 638 74
XM_006710386.3 XP_006710449.1 2 - 3 1676 74
XM_017000448.1 XP_016855937.1 2 - 3 1676 74
GenBank mRNA AK304611.1 - 2 - 3 557 74

View Full Product Details