The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114132.1 | NP_001107604.1 | 18 - 19 | 117 | |
XM_005246787.3 | XP_005246844.1 | 18 - 19 | 117 | ||
XM_006712698.3 | XP_006712761.1 | 19 - 20 | 117 | ||
XM_006712699.3 | XP_006712762.1 | 19 - 20 | 117 | ||
XM_011511658.2 | XP_011509960.1 | 19 - 20 | 117 | ||
XM_011511659.2 | XP_011509961.1 | 19 - 20 | 117 | ||
XM_011511660.2 | XP_011509962.1 | 15 - 16 | 117 | ||
XM_011511662.2 | XP_011509964.1 | 11 - 12 | 117 | ||
XM_011511663.2 | XP_011509965.1 | 19 - 20 | 117 | ||
XM_017004728.1 | XP_016860217.1 | 6 - 7 | 117 | ||
GenBank mRNA | DV080456.1 | - | 2 - 3 | 287 | 117 |