Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LDLRAD1
Entrez Gene ID
388633
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 54007298 - 54018186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010978.3 NP_001010978.2 2 - 3 150 99
GenBank mRNA BC127708.1 - 2 - 3 147 99
CF887342.1 - 2 - 3 150 99

View Full Product Details